Assay Details
| Gene Symbol: MED20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305455.1 | NP_001292384.1 | 2 - 3 | 390 | 83 | |
| NM_001305456.1 | NP_001292385.1 | 4 - 5 | 662 | 83 | ||
| NM_004275.4 | NP_004266.2 | 3 - 4 | 545 | 83 | ||
| GenBank mRNA | AJ420544.1 | - | 2 - 3 | 348 | 83 | |
| AK023092.1 | - | 3 - 4 | 489 | 83 | ||
| AK097385.1 | - | 4 - 5 | 662 | 83 | ||
| AK300573.1 | - | 2 - 3 | 382 | 83 | ||
| AL050196.1 | - | 1 - 2 | 60 | 83 | ||
| BC012618.1 | - | 3 - 4 | 541 | 4272961 | 83 | |
| BC015189.1 | - | 1 - 2 | 59 | 83 | ||
| BC019866.1 | - | 3 - 4 | 522 | 4470496 | 83 | |
| BC032552.1 | - | 3 - 4 | 507 | 5555309 | 83 | |
| BC040950.1 | - | 3 - 4 | 481 | 6042296 | 83 | |
More Information
Set Membership: |
|

MED20
Genomic Map