Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003958.3 | NP_003949.1 | 4 - 5 | 1234 | 78 | |
| NM_183078.2 | NP_898901.1 | 4 - 5 | 1234 | 78 | ||
| NR_046399.1 | 4 - 5 | 1340 | 78 | |||
| XM_017011462.1 | XP_016866951.1 | 4 - 5 | 1085 | 78 | ||
| XM_017011463.1 | XP_016866952.1 | 4 - 5 | 1090 | 78 | ||
| XM_017011464.1 | XP_016866953.1 | 4 - 5 | 1239 | 78 | ||
| GenBank mRNA | AB014546.1 | - | 4 - 5 | 1153 | 78 | |
| AF334675.1 | - | 4 - 5 | 1041 | 78 | ||
| AK022075.1 | - | 4 - 5 | 1135 | 78 | ||
| AK298319.1 | - | 4 - 5 | 1258 | 78 | ||
| BC007517.2 | - | 4 - 5 | 1163 | 3028746 | 78 | |
| BT007446.1 | - | 4 - 5 | 1041 | 78 | ||
More Information
Set Membership: |
|

RNF8
Genomic Map