Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003958.3 | NP_003949.1 | 5 - 6 | 1320 | 78 | |
| NM_183078.2 | NP_898901.1 | 5 - 6 | 1320 | 78 | ||
| NR_046399.1 | 5 - 6 | 1426 | 78 | |||
| XM_017011462.1 | XP_016866951.1 | 5 - 6 | 1171 | 78 | ||
| XM_017011463.1 | XP_016866952.1 | 5 - 6 | 1176 | 78 | ||
| XM_017011464.1 | XP_016866953.1 | 5 - 6 | 1325 | 78 | ||
| GenBank mRNA | AB014546.1 | - | 5 - 6 | 1239 | 78 | |
| AF334675.1 | - | 5 - 6 | 1127 | 78 | ||
| AK022075.1 | - | 5 - 6 | 1221 | 78 | ||
| AK222765.1 | - | 5 - 6 | 1238 | 78 | ||
| AK298319.1 | - | 5 - 6 | 1344 | 78 | ||
| BC007517.2 | - | 5 - 6 | 1249 | 3028746 | 78 | |
| BG029939.1 | - | 3 - 4 | 353 | 78 | ||
| BT007446.1 | - | 5 - 6 | 1127 | 78 | ||
| CA418096.1 | - | 2 - 3 | 618 | 78 | ||
More Information
Set Membership: |
|

RNF8
Genomic Map