Assay Details
| Gene Symbol: MRPS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032476.3 | NP_115865.1 | 1 - 2 | 223 | 76 | |
| GenBank mRNA | AB049942.1 | - | 1 - 2 | 169 | 76 | |
| AK312084.1 | - | 1 - 2 | 168 | 76 | ||
| AY061855.1 | - | 1 - 2 | 223 | 76 | ||
| BC000547.2 | - | 1 - 2 | 155 | 3162515 | 76 | |
| BC004976.1 | - | 1 - 2 | 157 | 2958115 | 76 | |
| BC010076.2 | - | 1 - 2 | 157 | 4304420 | 76 | |
| BC051302.1 | - | 1 - 2 | 64 | 4855355 | 76 | |
| BG720377.1 | - | 1 - 2 | 173 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000134.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008259.hg.1 | ENST00000381143.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001124.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001124.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001124.hg.1 | TC21000134.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001143.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001143.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001143.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001143.hg.1 | TC21000134.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001129.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001129.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001129.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001129.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001129.hg.1 | TC21000134.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013379.hg.1 | ENST00000603670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001130.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001130.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001130.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001130.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001130.hg.1 | TC21000134.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | TC2100008496.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157936.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008259.hg.1 | ENST00000381143.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013379.hg.1 | ENST00000603670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008496.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157932.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157932.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157932.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157932.hg.1 | TC2100008496.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100157932.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157932.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157932.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | TC2100008496.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000482679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000483977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | ENST00000488492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157953.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | NM_032476.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | TC2100008496.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | ENST00000381151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | ENST00000399312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | ENST00000477091.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | NM_006933.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157938.hg.1 | NM_032476.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 143207 | - | - |

MRPS6
Genomic Map