Assay Details
| Gene Symbol: YWHAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011517290.2 | XP_011515592.1 | 2 - 3 | 1008 | 77 | |
| XM_017013811.1 | XP_016869300.1 | 2 - 3 | 309 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017013811.1 transcripts.
Set Membership: |
|

YWHAZ
Genomic Map