Assay Details
| Gene Symbol: GIGYF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022574.4 | NP_072096.2 | 2 - 3 | 1172 | 58 | |
| XM_005250532.1 | XP_005250589.1 | 4 - 5 | 953 | 58 | ||
| XM_011516472.1 | XP_011514774.1 | 4 - 5 | 953 | 58 | ||
| XM_011516477.1 | XP_011514779.1 | 4 - 5 | 953 | 58 | ||
| XM_017012526.1 | XP_016868015.1 | 4 - 5 | 953 | 58 | ||
| XM_017012527.1 | XP_016868016.1 | 6 - 7 | 851 | 58 | ||
| XM_017012528.1 | XP_016868017.1 | 5 - 6 | 463 | 58 | ||
| XM_017012529.1 | XP_016868018.1 | 5 - 6 | 470 | 58 | ||
| XM_017012530.1 | XP_016868019.1 | 5 - 6 | 724 | 58 | ||
| GenBank mRNA | AY176044.1 | - | 2 - 3 | 1373 | 58 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001676.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025906.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025906.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025906.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025906.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025910.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025910.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025910.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025911.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025911.hg.1 | ENST00000539430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025911.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025911.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025905.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025905.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025905.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025905.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025907.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025907.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025907.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025907.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025912.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025912.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025912.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700186346.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186346.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186346.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186346.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186346.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186346.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186346.hg.1 | TC0700012013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700186348.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186348.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186348.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186348.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186348.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186348.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186348.hg.1 | TC0700012013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700186347.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186347.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186347.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186347.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186347.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186347.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186347.hg.1 | TC0700012013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186350.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186350.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186350.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186350.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186350.hg.1 | TC0700012013.hg.1 | Clariom D TC |

GIGYF1
Genomic Map