Assay Details
| Gene Symbol: GIGYF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022574.4 | NP_072096.2 | 5 - 6 | 1493 | 133 | |
| XM_005250532.1 | XP_005250589.1 | 7 - 8 | 1274 | 133 | ||
| XM_011516472.1 | XP_011514774.1 | 7 - 8 | 1274 | 133 | ||
| XM_011516477.1 | XP_011514779.1 | 7 - 8 | 1274 | 133 | ||
| XM_017012526.1 | XP_016868015.1 | 7 - 8 | 1274 | 133 | ||
| XM_017012527.1 | XP_016868016.1 | 9 - 10 | 1172 | 133 | ||
| XM_017012528.1 | XP_016868017.1 | 8 - 9 | 784 | 133 | ||
| XM_017012529.1 | XP_016868018.1 | 8 - 9 | 791 | 133 | ||
| XM_017012530.1 | XP_016868019.1 | 8 - 9 | 1045 | 133 | ||
| XM_017012531.1 | XP_016868020.1 | 3 - 4 | 297 | 133 | ||
| GenBank mRNA | AY176044.1 | - | 5 - 6 | 1694 | 133 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001676.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07025899.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025899.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025899.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025900.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025900.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025900.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025901.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025901.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025901.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186341.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186341.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186341.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186341.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186341.hg.1 | TC0700012013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700186340.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186340.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186340.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186340.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186340.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186340.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186340.hg.1 | TC0700012013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186342.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186342.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186342.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186342.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186342.hg.1 | TC0700012013.hg.1 | Clariom D TC |

GIGYF1
Genomic Map