Assay Details
| Gene Symbol: GABRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000809.3 | NP_000800.2 | 6 - 7 | 1703 | 74 | |
| NM_001204266.1 | NP_001191195.1 | 6 - 7 | 733 | 74 | ||
| GenBank mRNA | AK090780.1 | - | 6 - 7 | 1703 | 74 | |
| BC035055.1 | - | 6 - 7 | 832 | 5259426 | 74 | |
| CD013893.1 | - | 6 - 7 | 737 | 74 | ||
| KJ534840.1 | - | 6 - 7 | 720 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001165.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
gamma-aminobutyric acid signaling pathway
central nervous system development
ion transmembrane transport
chloride transmembrane transport
regulation of response to drug
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
benzodiazepine receptor activity
transport
gamma-aminobutyric acid signaling pathway
central nervous system development
ion transmembrane transport
chloride transmembrane transport
regulation of response to drug
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
benzodiazepine receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04015984.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04015984.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04015984.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04015984.hg.1 | TC04001165.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | ENST00000264318.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | ENST00000502874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | ENST00000508560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | NM_000809.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | NM_001204266.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172248.hg.1 | TC0400010602.hg.1 | Clariom D TC |

GABRA4
Genomic Map