| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001346.hg.1 |
ENST00000314103.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001346.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001346.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001346.hg.1 |
TC21000154.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001349.hg.1 |
ENST00000314103.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001349.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001349.hg.1 |
TC21000154.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001350.hg.1 |
ENST00000314103.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001350.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001350.hg.1 |
TC21000154.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100158114.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158114.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158114.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100158114.hg.1 |
TC2100008508.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100158114.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158114.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158114.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000480486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008508.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100158119.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158119.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158119.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100158119.hg.1 |
TC2100008508.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100158119.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158119.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158119.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100158122.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158122.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158122.hg.1 |
NM_005441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100158122.hg.1 |
TC2100008508.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100158122.hg.1 |
ENST00000314103.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158122.hg.1 |
ENST00000481458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158122.hg.1 |
NM_005441.2 |
Refseq |