Assay Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031745.4 | NP_001026915.1 | 2 - 3 | 229 | 81 | |
| NM_001267053.3 | NP_001253982.1 | 2 - 3 | 229 | 81 | ||
| NM_144968.3 | NP_659405.1 | 2 - 3 | 229 | 81 | ||
| XM_005261988.3 | XP_005262045.1 | 2 - 3 | 437 | 81 | ||
| XM_005261990.3 | XP_005262047.1 | 2 - 3 | 303 | 81 | ||
| GenBank mRNA | AK057345.1 | - | 2 - 3 | 195 | 81 | |
| AK290346.1 | - | 2 - 3 | 179 | 81 | ||
| AK292871.1 | - | 2 - 3 | 184 | 81 | ||
| AK304179.1 | - | 2 - 3 | 229 | 81 | ||
| BC036767.1 | - | 2 - 3 | 144 | 5744127 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000316.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK057345.1,AK290346.1,AK292871.1,AK304179.1,BC036767.1,NM_001031745.4,NM_001267053.3,NM_144968.3,XM_005261988.3,XM_005261990.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004180.hg.1 | ENST00000375327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004180.hg.1 | ENST00000414955.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004180.hg.1 | ENST00000457095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004180.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004180.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004180.hg.1 | TC0X000316.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150407.hg.1 | TC0X00007325.hg.1 | Clariom D TC |

RIBC1
Genomic Map