Assay Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031745.4 | NP_001026915.1 | 6 - 7 | 899 | 90 | |
| NM_001267053.3 | NP_001253982.1 | 5 - 6 | 554 | 90 | ||
| XM_005261988.3 | XP_005262045.1 | 6 - 7 | 1107 | 90 | ||
| XM_005261990.3 | XP_005262047.1 | 5 - 6 | 628 | 90 | ||
| GenBank mRNA | AK292871.1 | - | 6 - 7 | 854 | 90 | |
| AK304179.1 | - | 5 - 6 | 554 | 90 | ||
| BC036767.1 | - | 6 - 7 | 814 | 5744127 | 90 | |
| BM978850.1 | - | 2 - 3 | 90 | |||
| CF272701.1 | - | 1 - 2 | 90 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150416.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150416.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150416.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150416.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150416.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150416.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150416.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150416.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150416.hg.1 | TC0X00007325.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00150418.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150418.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150418.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150418.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150418.hg.1 | TC0X00007325.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |

RIBC1
Genomic Map