Assay Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031745.4 | NP_001026915.1 | 7 - 8 | 74 | ||
| XM_005261988.3 | XP_005262045.1 | 7 - 8 | 74 | |||
| XM_005261990.3 | XP_005262047.1 | 6 - 7 | 74 | |||
| GenBank mRNA | AI127598.1 | - | 1 - 2 | 207 | 74 | |
| AK292871.1 | - | 7 - 8 | 74 | |||
| BC036767.1 | - | 7 - 8 | 5744127 | 74 | ||
| BM978850.1 | - | 1 - 2 | 225 | 74 | ||
| CA433728.1 | - | 1 - 2 | 219 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000316.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004190.hg.1 | ENST00000375327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004190.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004190.hg.1 | TC0X000316.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00150420.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150420.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150420.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150420.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150420.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150420.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150420.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150420.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150420.hg.1 | TC0X00007325.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150419.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150419.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150419.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150419.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150419.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150419.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150419.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150419.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150419.hg.1 | TC0X00007325.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 168429 | - | - |

RIBC1
Genomic Map