| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028170.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028170.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028170.hg.1 |
TC06002240.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028164.hg.1 |
ENST00000229758.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028164.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028164.hg.1 |
ENST00000477822.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028164.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028164.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028164.hg.1 |
TC06002240.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028171.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06028171.hg.1 |
TC06002240.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600194047.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600194047.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600194047.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600194047.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600194047.hg.1 |
TC0600013602.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013602.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600194048.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600194048.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600194048.hg.1 |
TC0600013602.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
ENST00000229758.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
ENST00000367241.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
ENST00000477822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
NM_001142522.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
NM_012177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600194039.hg.1 |
TC0600013602.hg.1 |
Clariom D TC |