Assay Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142522.1 | NP_001135994.1 | 3 - 4 | 1176 | 115 | |
| NM_012177.3 | NP_036309.1 | 3 - 4 | 1016 | 115 | ||
| GenBank mRNA | AF129535.1 | - | 3 - 4 | 968 | 115 | |
| AJ420499.1 | - | 3 - 4 | 1036 | 115 | ||
| AK026197.1 | - | 4 - 5 | 925 | 115 | ||
| AK055221.1 | - | 3 - 4 | 1150 | 115 | ||
| AY079515.1 | - | 3 - 4 | 907 | 115 | ||
| BC018905.1 | - | 3 - 4 | 958 | 3960052 | 115 | |
More Information
Set Membership: |
|

FBXO5
Genomic Map