| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024952.hg.1 |
ENST00000274721.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024952.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024952.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024952.hg.1 |
TC05001828.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500184330.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184330.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184330.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184330.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184330.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184330.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184330.hg.1 |
TC0500012164.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |