Assay Details
| Gene Symbol: HPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002150.2 | NP_002141.1 | 2 - 3 | 68 | 94 | |
| GenBank mRNA | AK290826.1 | - | 2 - 3 | 64 | 94 | |
| CD675915.1 | - | 2 - 3 | 197 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002066.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290826.1,NM_002150.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027170.hg.1 | ENST00000289004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027170.hg.1 | ENST00000543163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027170.hg.1 | ENST00000545969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027170.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027170.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027170.hg.1 | TC12002066.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027171.hg.1 | ENST00000289004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027171.hg.1 | ENST00000543163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027171.hg.1 | ENST00000545969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027171.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027171.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027171.hg.1 | TC12002066.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195483.hg.1 | TC1200012215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |

HPD
Genomic Map