Assay Details
| Gene Symbol: ITPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002223.3 | NP_002214.2 | 27 - 28 | 3970 | 108 | |
| XM_017019266.1 | XP_016874755.1 | 28 - 29 | 4029 | 108 | ||
| XM_017019267.1 | XP_016874756.1 | 28 - 29 | 3553 | 108 | ||
| XM_017019268.1 | XP_016874757.1 | 27 - 28 | 3430 | 108 | ||
| XM_017019269.1 | XP_016874758.1 | 28 - 29 | 4029 | 108 | ||
| GenBank mRNA | BU173910.1 | - | 2 - 3 | 255 | 108 | |
| D26350.1 | - | 27 - 28 | 3971 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001328.hg.1
Set Membership: |
|

ITPR2
Genomic Map