Assay Details
| Gene Symbol: ITPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002223.3 | NP_002214.2 | 36 - 37 | 5382 | 143 | |
| XM_017019266.1 | XP_016874755.1 | 37 - 38 | 5441 | 143 | ||
| XM_017019267.1 | XP_016874756.1 | 37 - 38 | 4965 | 143 | ||
| XM_017019268.1 | XP_016874757.1 | 36 - 37 | 4842 | 143 | ||
| GenBank mRNA | D26350.1 | - | 36 - 37 | 5383 | 143 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001328.hg.1
Set Membership: |
|

ITPR2
Genomic Map