Assay Details
| Gene Symbol: ITPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002223.3 | NP_002214.2 | 46 - 47 | 6866 | 153 | |
| XM_017019266.1 | XP_016874755.1 | 47 - 48 | 6925 | 153 | ||
| XM_017019267.1 | XP_016874756.1 | 47 - 48 | 6449 | 153 | ||
| XM_017019268.1 | XP_016874757.1 | 46 - 47 | 6326 | 153 | ||
| GenBank mRNA | AK307686.1 | - | 9 - 10 | 1277 | 153 | |
| D26350.1 | - | 46 - 47 | 6867 | 153 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001328.hg.1
Set Membership: |
|

ITPR2
Genomic Map