Assay Details
| Gene Symbol: GPR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005295.2 | NP_005286.2 | 2 - 3 | 1318 | 113 | |
| GenBank mRNA | AK122621.1 | - | 2 - 3 | 1315 | 113 | |
| AK307393.1 | - | 2 - 3 | 1315 | 113 | ||
| BX647649.1 | - | 1 - 2 | 182 | 113 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000691.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK122621.1,NM_005295.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07010801.hg.1 | TC07000691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010801.hg.1 | ENST00000304402.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010801.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010802.hg.1 | TC07000691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010802.hg.1 | ENST00000304402.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010802.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010796.hg.1 | TC07000691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010796.hg.1 | ENST00000304402.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010796.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010796.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010798.hg.1 | TC07000691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010798.hg.1 | ENST00000304402.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010798.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010798.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010797.hg.1 | TC07000691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010797.hg.1 | ENST00000304402.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010797.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010797.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010795.hg.1 | TC07000691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010795.hg.1 | ENST00000304402.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010795.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010799.hg.1 | TC07000691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010799.hg.1 | ENST00000304402.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010799.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161808.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161808.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161808.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161808.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161808.hg.1 | TC0700008753.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161810.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161810.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161810.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161810.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161810.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161810.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161810.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161810.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161810.hg.1 | TC0700008753.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161809.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161809.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161809.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161809.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161809.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161809.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161809.hg.1 | TC0700008753.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161815.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161815.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161815.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161815.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161815.hg.1 | TC0700008753.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161812.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161812.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161812.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161812.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161812.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161812.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161812.hg.1 | TC0700008753.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161814.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161814.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161814.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161814.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161814.hg.1 | TC0700008753.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008753.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161811.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161811.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161811.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161811.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161811.hg.1 | ENST00000304402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161811.hg.1 | ENST00000473300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161811.hg.1 | ENST00000496754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161811.hg.1 | NM_005295.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161811.hg.1 | TC0700008753.hg.1 | Clariom D TC |

GPR22
Genomic Map