Assay Details
| Gene Symbol: PITRM1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038284.1 | 1 - 2 | 170 | 78 | ||
| GenBank mRNA | BC050332.1 | - | 1 - 2 | 99 | 78 | |
| DB040375.1 | - | 1 - 2 | 170 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000023.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10000241.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000241.hg.1 | ENST00000452662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000241.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000241.hg.1 | TC10000023.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000238.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000238.hg.1 | TC10000023.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000240.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000240.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000240.hg.1 | TC10000023.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000239.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000239.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000239.hg.1 | TC10000023.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000430356.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000430356.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184970.hg.1 | TC1000012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | ENST00000430356.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | ENST00000430356.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184971.hg.1 | TC1000012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000430356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012424.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | ENST00000598280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184969.hg.1 | TC1000012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000184968.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184968.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184968.hg.1 | TC1000012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | ENST00000430356.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | ENST00000430356.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | ENST00000441377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | ENST00000601046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | NR_038284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184972.hg.1 | TC1000012424.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 125054 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 233166 | - | - |

PITRM1-AS1
Genomic Map