Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | poly(A) binding protein cytoplasmic 3 | PABPC3 |
PABP3, PABPL3, tPABP |
-43.0kb | Sense | Chr. 13 - 25096138 - 25098567 on NCBI Build GRCh38 |
| Downstream | APC membrane recruitment protein 2 | AMER2 | FAM123A | 19.9kb | Anti-sense | Chr. 13 - 25161679 - 25172456 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map