Assay Details
| Gene Symbol: PDE6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006204.3 | NP_006195.3 | 3 - 4 | 857 | 108 | |
| GenBank mRNA | U31973.1 | - | 3 - 4 | 857 | 108 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000667.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000475427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000475427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000475427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | ENST00000475427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007508.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007508.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007508.hg.1 | TC10000667.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000475427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000475427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000475427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | ENST00000475427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008488.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000157710.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000157710.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000157710.hg.1 | ENST00000371447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000157710.hg.1 | NM_006204.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000157710.hg.1 | TC1000008488.hg.1 | Clariom D TC |

PDE6C
Genomic Map