Assay Details
| Gene Symbol: PDE6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006204.3 | NP_006195.3 | 8 - 9 | 1256 | 129 | |
| XM_017016322.1 | XP_016871811.1 | 6 - 7 | 620 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000667.hg.1
Set Membership: |
|

PDE6C
Genomic Map