Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adhesion G protein-coupled receptor G6 | ADGRG6 |
APG1, DREG, GPR126, LCCS9, PS1TP2, VIGR |
Anti-sense | Chr. 6 - 142301919 - 142446266 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC153910 | LOC153910 | 160.8kb | Sense | Chr. 6 - 142526455 - 142637889 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map