Assay Details
| Gene Symbol: NR2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286102.1 | NP_001273031.1 | 3 - 4 | 378 | 113 | |
| NM_003269.4 | NP_003260.1 | 3 - 4 | 971 | 113 | ||
| GenBank mRNA | AB307709.1 | - | 3 - 4 | 263 | 113 | |
| AF220532.1 | - | 3 - 4 | 970 | 113 | ||
| AF411525.1 | - | 3 - 4 | 504 | 113 | ||
| AK131541.1 | - | 3 - 4 | 378 | 113 | ||
| BC028031.1 | - | 3 - 4 | 468 | 5242079 | 113 | |
| HQ692846.1 | - | 3 - 4 | 263 | 113 | ||
| Y13276.1 | - | 3 - 4 | 431 | 113 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000860.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
behavioral fear response
aggressive behavior
transcription initiation from RNA polymerase II promoter
nervous system development
visual perception
dentate gyrus development
amygdala development
olfactory bulb development
layer formation in cerebral cortex
forebrain generation of neurons
cerebral cortex neuron differentiation
anterior commissure morphogenesis
extracellular matrix organization
somatic stem cell population maintenance
social behavior
negative regulation of apoptotic process
steroid hormone mediated signaling pathway
cell fate commitment
negative regulation of neuron differentiation
positive regulation of angiogenesis
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
negative regulation of astrocyte differentiation
regulation of dendrite morphogenesis
retina development in camera-type eye
regulation of timing of neuron differentiation
long-term synaptic potentiation
regulation of cell migration involved in sprouting angiogenesis
negative regulation of neural precursor cell proliferation
positive regulation of neural precursor cell proliferation
positive regulation of stem cell proliferation
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
steroid hormone receptor activity
zinc ion binding
histone deacetylase binding
sequence-specific DNA binding
negative regulation of transcription from RNA polymerase II promoter
behavioral fear response
aggressive behavior
transcription initiation from RNA polymerase II promoter
nervous system development
visual perception
dentate gyrus development
amygdala development
olfactory bulb development
layer formation in cerebral cortex
forebrain generation of neurons
cerebral cortex neuron differentiation
anterior commissure morphogenesis
extracellular matrix organization
somatic stem cell population maintenance
social behavior
negative regulation of apoptotic process
steroid hormone mediated signaling pathway
cell fate commitment
negative regulation of neuron differentiation
positive regulation of angiogenesis
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
negative regulation of astrocyte differentiation
regulation of dendrite morphogenesis
retina development in camera-type eye
regulation of timing of neuron differentiation
long-term synaptic potentiation
regulation of cell migration involved in sprouting angiogenesis
negative regulation of neural precursor cell proliferation
positive regulation of neural precursor cell proliferation
positive regulation of stem cell proliferation
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
steroid hormone receptor activity
zinc ion binding
histone deacetylase binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010092.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010092.hg.1 | ENST00000368986.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010092.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010092.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010092.hg.1 | TC06000860.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162135.hg.1 | TC0600009041.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000368986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000426403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | ENST00000484978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_001286102.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009041.hg.1 | NM_003269.4 | Refseq |

NR2E1
Genomic Map