| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010094.hg.1 |
ENST00000368983.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010094.hg.1 |
ENST00000368986.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010094.hg.1 |
ENST00000484978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010094.hg.1 |
NM_003269.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010094.hg.1 |
TC06000860.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010093.hg.1 |
ENST00000368983.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010093.hg.1 |
ENST00000368986.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010093.hg.1 |
ENST00000484978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010093.hg.1 |
NM_003269.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010093.hg.1 |
TC06000860.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
ENST00000368983.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
ENST00000368986.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
ENST00000484978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
NM_001286102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
NM_003269.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
ENST00000368983.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
ENST00000368986.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
ENST00000484978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
NM_001286102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
NM_003269.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162136.hg.1 |
TC0600009041.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600162138.hg.1 |
ENST00000368983.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162138.hg.1 |
ENST00000368986.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162138.hg.1 |
NM_001286102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162138.hg.1 |
NM_003269.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162138.hg.1 |
ENST00000368983.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162138.hg.1 |
ENST00000368986.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162138.hg.1 |
NM_001286102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162138.hg.1 |
NM_003269.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162138.hg.1 |
TC0600009041.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
ENST00000368983.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
ENST00000368986.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
ENST00000484978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
NM_001286102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
NM_003269.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
ENST00000368983.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
ENST00000368986.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
ENST00000484978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
NM_001286102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
NM_003269.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162137.hg.1 |
TC0600009041.hg.1 |
Clariom D TC |