| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008696.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008696.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008696.hg.1 |
TC04000598.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008697.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008697.hg.1 |
ENST00000433996.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008697.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008697.hg.1 |
TC04000598.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158500.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400158500.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158500.hg.1 |
ENST00000296499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400158500.hg.1 |
NM_004784.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158500.hg.1 |
TC0400008534.hg.1 |
Clariom D TC |