| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009860.hg.1 |
ENST00000369214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009860.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009860.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009860.hg.1 |
TC06000831.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009861.hg.1 |
ENST00000369214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009861.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009861.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009861.hg.1 |
TC06000831.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009864.hg.1 |
ENST00000369214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009864.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009864.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009864.hg.1 |
TC06000831.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600161603.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161603.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161603.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161603.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161603.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161603.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161603.hg.1 |
TC0600008912.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008912.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161601.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161601.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161601.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161601.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161601.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161601.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161601.hg.1 |
TC0600008912.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600161600.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161600.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161600.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161600.hg.1 |
ENST00000369214.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161600.hg.1 |
ENST00000369215.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161600.hg.1 |
NM_021620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161600.hg.1 |
TC0600008912.hg.1 |
Clariom D TC |