Assay Details
| Gene Symbol: SASS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304829.1 | NP_001291758.1 | 10 - 11 | 1532 | 176 | |
| NM_194292.2 | NP_919268.1 | 11 - 12 | 1612 | 176 | ||
| XM_017000486.1 | XP_016855975.1 | 11 - 12 | 1378 | 176 | ||
| GenBank mRNA | AK302513.1 | - | 12 - 13 | 1362 | 176 | |
| AK307824.1 | - | 10 - 11 | 1398 | 176 | ||
| AL834265.1 | - | 7 - 8 | 875 | 176 | ||
| AY359522.1 | - | 11 - 12 | 1472 | 176 | ||
| BC101026.3 | - | 10 - 11 | 1325 | 40007432 | 176 | |
| BX648406.1 | - | 11 - 12 | 1369 | 176 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002916.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045310.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045310.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045310.hg.1 | ENST00000535161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045310.hg.1 | ENST00000539329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045310.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045310.hg.1 | TC01002916.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100212163.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212163.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212163.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212163.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212163.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212163.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212163.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212163.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212163.hg.1 | TC0100015058.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |

SASS6
Genomic Map