Assay Details
| Gene Symbol: CR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY114160.1 | - | 10 - 11 | 1476 | 74 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000430248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000430248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000430248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000430248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000294997.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000430248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000430248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000430248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000430248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000441364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000508064.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000530905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | ENST00000531844.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011417.hg.1 | NM_175710.1 | Refseq |

CR1L
Genomic Map