Assay Details
| Gene Symbol: ZNF704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033723.2 | NP_001028895.1 | 3 - 4 | 562 | 72 | |
| XM_005251279.4 | XP_005251336.2 | 3 - 4 | 1193 | 72 | ||
| XM_017013725.1 | XP_016869214.1 | 3 - 4 | 609 | 72 | ||
| GenBank mRNA | AK131274.1 | - | 3 - 4 | 562 | 72 | |
| BC136844.1 | - | 3 - 4 | 375 | 9020835 | 72 | |
| BC136845.1 | - | 3 - 4 | 375 | 9020836 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001359.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000517379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000517379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000517986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000517986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017547.hg.1 | ENST00000327835.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017547.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017547.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017547.hg.1 | TC08001359.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017550.hg.1 | ENST00000327835.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017550.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017550.hg.1 | TC08001359.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017546.hg.1 | ENST00000327835.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017546.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017546.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017546.hg.1 | TC08001359.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017553.hg.1 | ENST00000327835.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017553.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017553.hg.1 | TC08001359.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800174190.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174190.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174190.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174190.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174190.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174190.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174190.hg.1 | TC0800010918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800174194.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174194.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174194.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174194.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174194.hg.1 | TC0800010918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800174186.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174186.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174186.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174186.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174186.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174186.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174186.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174186.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174186.hg.1 | TC0800010918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800174187.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174187.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174187.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174187.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174187.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174187.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174187.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174187.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174187.hg.1 | TC0800010918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000517379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000517379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000517986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000517986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |

ZNF704
Genomic Map