| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017541.hg.1 |
ENST00000327835.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017541.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017541.hg.1 |
TC08001359.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000517379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000517379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000517986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000517986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000519936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000519936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000519936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000519936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000522040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000522040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
ENST00000522040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000517379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000517379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000517986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000517986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000519936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000519936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000519936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000519936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000520336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000522040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000522040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
ENST00000522040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010918.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800174181.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174181.hg.1 |
ENST00000522040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174181.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800174181.hg.1 |
ENST00000327835.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174181.hg.1 |
ENST00000522040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174181.hg.1 |
NM_001033723.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800174181.hg.1 |
TC0800010918.hg.1 |
Clariom D TC |