Assay Details
Gene Symbol: DISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020780.1 | NP_065831.1 | 16 - 17 | 3369 | 103 | |
XM_011541828.2 | XP_011540130.1 | 15 - 16 | 5471 | 103 | ||
XM_011541829.1 | XP_011540131.1 | 15 - 16 | 2344 | 103 | ||
GenBank mRNA | AB037758.1 | - | 16 - 17 | 3369 | 103 | |
AB593117.1 | - | 16 - 17 | 3452 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000142.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
smoothened signaling pathway
cholesterol metabolic process
regulation of lipid transport
cholesterol homeostasis
negative regulation of neuron differentiation
positive regulation of lipid metabolic process
positive regulation of neural precursor cell proliferation
molecular_function
smoothened signaling pathway
cholesterol metabolic process
regulation of lipid transport
cholesterol homeostasis
negative regulation of neuron differentiation
positive regulation of lipid metabolic process
positive regulation of neural precursor cell proliferation
molecular_function
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000423056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000423056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | ENST00000423056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01002225.hg.1 | TC01000142.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01002225.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01002225.hg.1 | ENST00000389575.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01002225.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000304391.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000423056.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000423056.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | ENST00000423056.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006857.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100147544.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100147544.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100147544.hg.1 | ENST00000294484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100147544.hg.1 | NM_020780.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100147544.hg.1 | TC0100006857.hg.1 | Clariom D TC |
