Assay Details
| Gene Symbol: MOXD2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024346.3 | 2 - 3 | 436 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001936.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030087.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030087.hg.1 | TC07001936.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192692.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192692.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192692.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192692.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192692.hg.1 | TC0700012854.hg.1 | Clariom D TC |

MOXD2P
Genomic Map