Assay Details
| Gene Symbol: MOXD2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024346.3 | 3 - 4 | 602 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001936.hg.1
Set Membership: |
|

MOXD2P
Genomic Map