Assay Details
| Gene Symbol: MOXD2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024346.3 | 6 - 7 | 948 | 153 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001936.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 197379 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 208567 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3263629 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4515 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4516 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17127 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P25813856 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P25813856 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3301778 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3263629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835332 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094336 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835368 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835345 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094318 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094318 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835345 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094336 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835368 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26094320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05835332 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835332 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835371 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16025878 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835263 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835274 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835271 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835281 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835340 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835377 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835339 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835330 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835381 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835297 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835382 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835367 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835291 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835260 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835351 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835379 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835256 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835315 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835335 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835325 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835328 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835251 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835301 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835272 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835347 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835299 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835308 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835295 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835369 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16025877 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835317 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835311 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835269 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835338 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835313 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835265 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835349 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835372 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835249 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835343 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835337 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835267 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835280 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835303 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835250 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835277 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835364 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835342 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835254 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16025875 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05835284 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18144537 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07030082.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030082.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030082.hg.1 | TC07001936.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192683.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192683.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192683.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192683.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192683.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192683.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192683.hg.1 | TC0700012854.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007291 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007291 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3236116 | NR_024346.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236116 | DY655576.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236116 | NR_024346.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236116 | DY655576.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236116 | NR_024346.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236116 | DY655576.1 | human_transcriptome |

MOXD2P
Genomic Map