Assay Details
| Gene Symbol: TMEM235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204210.1 | NP_001191139.1 | 4 - 5 | 1029 | 77 | |
| NM_001204211.1 | NP_001191140.1 | 3 - 4 | 948 | 77 | ||
| XM_011524624.2 | XP_011522926.1 | 3 - 4 | 405 | 77 | ||
| GenBank mRNA | BI757060.1 | - | 3 - 4 | 367 | 77 | |
| BI757981.1 | - | 3 - 4 | 299 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000900.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17011720.hg.1 | ENST00000374946.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011720.hg.1 | ENST00000421688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011720.hg.1 | ENST00000543327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011720.hg.1 | ENST00000550981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011720.hg.1 | ENST00000551068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011720.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011720.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011720.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011720.hg.1 | TC17000900.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | TC1700009044.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700168357.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000374946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000421688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000550981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000551068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000586400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | ENST00000591033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009044.hg.1 | NM_001204212.1 | Refseq |

TMEM235
Genomic Map