Assay Details
| Gene Symbol: SULT1C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320878.1 | NP_001307807.1 | 6 - 7 | 724 | 131 | |
| XM_017004153.1 | XP_016859642.1 | 5 - 6 | 1613 | 131 | ||
| XM_017004154.1 | XP_016859643.1 | 5 - 6 | 1613 | 131 | ||
More Information
Set Membership: |
|

SULT1C3
Genomic Map