Assay Details
| Gene Symbol: PCA3 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132312.1 | 2 - 3 | 281 | 80 | |||
| GenBank mRNA | AF103907.1 | - | LIT1552 DD3 |
2 - 3 | 281 | 80 | |
More Information
Set Membership: |
|

PCA3
Genomic Map