Assay Details
| Gene Symbol: TCAM1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002947.2 | 3 - 4 | 1151 | 91 | ||
| GenBank mRNA | AB026156.1 | - | 3 - 4 | 1151 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2 |
SMARCD2 |
BAF60B, CRACD2, PRO2451, Rsc6p |
-16.9kb | Anti-sense | Chr. 17 - 63832081 - 63842991 on NCBI Build GRCh38 |
| Downstream | chorionic somatomammotropin hormone 2 | CSH2 |
CS-2, CSB, GHB1, PL, hCS-B |
11.1kb | Anti-sense | Chr. 17 - 63872012 - 63873729 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000773.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000463377.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000463377.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000463377.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000466125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000466125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000466125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000466125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000476374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000476374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000476374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010102.hg.1 | ENST00000478379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010102.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010102.hg.1 | TC17000773.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | ENST00000476374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | TC1700008578.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | ENST00000476374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164221.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000463377.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000463377.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000463377.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000466125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000466125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000466125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000466125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000469014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000476374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000476374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000476374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008578.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700164222.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164222.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164222.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164222.hg.1 | NR_002947.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700164222.hg.1 | TC1700008578.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700164222.hg.1 | ENST00000478379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164222.hg.1 | ENST00000482005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164222.hg.1 | ENST00000485295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700164222.hg.1 | NR_002947.2 | Refseq |

TCAM1P
Genomic Map