| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029995.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029995.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029995.hg.1 |
ENST00000366615.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029995.hg.1 |
ENST00000424237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029995.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029995.hg.1 |
NM_001206641.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029995.hg.1 |
TC01001934.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029992.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029992.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029992.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029992.hg.1 |
TC01001934.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188647.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188647.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188647.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188647.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188647.hg.1 |
ENST00000366612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188647.hg.1 |
ENST00000366613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188647.hg.1 |
NM_001012985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188647.hg.1 |
NM_001301733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188647.hg.1 |
TC0100012020.hg.1 |
Clariom D TC |