| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011949.hg.1 |
ENST00000439341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011949.hg.1 |
ENST00000439341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011949.hg.1 |
ENST00000439341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011949.hg.1 |
NR_110681.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011949.hg.1 |
NR_110681.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011949.hg.1 |
NR_110681.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188148.hg.1 |
ENST00000439341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188148.hg.1 |
NR_110681.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188148.hg.1 |
ENST00000439341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188148.hg.1 |
NR_110681.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188148.hg.1 |
TC0100011949.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100011949.hg.1 |
ENST00000439341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011949.hg.1 |
ENST00000439341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011949.hg.1 |
ENST00000439341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011949.hg.1 |
NR_110681.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011949.hg.1 |
NR_110681.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011949.hg.1 |
NR_110681.1 |
Refseq |