Assay Details
| Gene Symbol: SNRPN | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF400487.1 | - | LIT2225 IC-SNURF-SNRPN 3 prime noncoding transcripts |
18 - 19 | 2484 | 81 | |
| AF400488.1 | - | LIT2213 IC-SNURF-SNRPN 3 prime noncoding transcripts |
12 - 13 | 1892 | 81 | ||
| AL832758.1 | - | 28 - 29 | 3717 | 81 | |||
| BC080646.1 | - | 28 - 29 | 3832 | 81 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 116-30 |
SNORD116-30 | HBII-85-30 | -1.2kb | Sense | Chr. 15 - 25108268 - 25108352 on NCBI Build GRCh38 |
| Downstream | imprinted in Prader-Willi syndrome (non-protein coding) | IPW | NCRNA00002 | 4.7kb | Sense | Chr. 15 - 25116545 - 25122476 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNRPN
Genomic Map