Assay Details
| Gene Symbol: SNRPN | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF400485.1 | - | LIT2216 IC-SNURF-SNRPN 3 prime noncoding transcripts |
4 - 5 | 462 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 116-1 |
SNORD116-1 |
HBII-85-1, PWCR1 |
Sense | Chr. 15 - 25051476 - 25051572 on NCBI Build GRCh38 | |
| Downstream |
small nucleolar RNA, C/D box 116-2 |
SNORD116-2 | HBII-85-2 | 293.0 | Sense | Chr. 15 - 25054209 - 25054305 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNRPN
Genomic Map