Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurotrimin | NTM |
HNT, IGLON2, NTRI |
-74.7kb | Anti-sense | Chr. 11 - 131370326 - 132336822 on NCBI Build GRCh38 |
| Downstream | opioid binding protein/cell adhesion molecule like | OPCML |
IGLON1, OBCAM, OPCM |
785.0 | Sense | Chr. 11 - 132403361 - 133532983 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map