| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012921.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012921.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012921.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012921.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012921.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012921.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193939.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193939.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193939.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193939.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193939.hg.1 |
TC0200012921.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200012921.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012921.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012921.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012921.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012921.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012921.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193940.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193940.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193940.hg.1 |
ENST00000457448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193940.hg.1 |
NR_110271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193940.hg.1 |
TC0200012921.hg.1 |
Clariom D TC |