Assay Details
| Gene Symbol: DUSP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017027654.1 | XP_016883143.1 | 7 - 8 | 815 | 66 | |
| XM_017027655.1 | XP_016883144.1 | 7 - 8 | 606 | 66 | ||
| XM_017027656.1 | XP_016883145.1 | 7 - 8 | 815 | 66 | ||
| XM_017027658.1 | XP_016883147.1 | 7 - 8 | 755 | 66 | ||
| XM_017027659.1 | XP_016883148.1 | 7 - 8 | 761 | 66 | ||
| XM_017027660.1 | XP_016883149.1 | 7 - 8 | 643 | 66 | ||
| XM_017027661.1 | XP_016883150.1 | 6 - 7 | 752 | 66 | ||
| XM_017027662.1 | XP_016883151.1 | 7 - 8 | 613 | 66 | ||
More Information
Set Membership: |
|

DUSP15
Genomic Map