Assay Details
| Gene Symbol: LOC107133515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015341.2 | 1 - 2 | 197 | 60 | ||
| GenBank mRNA | BG721507.1 | - | 1 - 2 | 158 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TBC1 domain family member 29 | TBC1D29 | -10.8kb | Sense | Chr. 17 - 30559566 - 30565667 on NCBI Build GRCh38 | |
| Downstream |
SH3 domain containing GRB2 like 1, endophilin A2 pseudogene 2 |
SH3GL1P2 |
CNSA-P2, SH3GLP2 |
24.1kb | Sense | Chr. 17 - 30624318 - 30626823 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000352.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398851.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398851.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398851.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398851.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398851.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000398851.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000412831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000412831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000412831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000412831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000412831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000412831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000412831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000412831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000417404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000431308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000440026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000514992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000514992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000514992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000514992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000514992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000514992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000578265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000579301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000579301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000579301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000579301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000579301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000579301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000579301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000583030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000583030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | ENST00000583030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012221.hg.1 | NR_015341.2 | Refseq |

LOC107133515
Genomic Map