| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002351.hg.1 |
ENST00000344962.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002351.hg.1 |
ENST00000409192.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002351.hg.1 |
ENST00000409653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002351.hg.1 |
ENST00000409994.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002351.hg.1 |
ENST00000483090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002351.hg.1 |
NM_001127364.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002351.hg.1 |
NM_001127365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002351.hg.1 |
NM_199136.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002351.hg.1 |
TC07000150.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000344962.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000409192.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000409653.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000409994.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000429719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000462546.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000483090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
NM_001127364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
NM_001127365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
NM_001300932.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
NM_199136.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000344962.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000409192.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000409653.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000409994.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000429719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000462546.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
ENST00000483090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
NM_001127364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
NM_001127365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
NM_001300932.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
NM_199136.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147956.hg.1 |
TC0700006931.hg.1 |
Clariom D TC |